Cyclohexyldiphenylphosphine oxide ≥98%
Supplier: Thermo Fisher Scientific
Warning
L00698.03 L00698.06
L00698.03EA 33.1 EUR
L00698.03 L00698.06
Cyclohexyldiphenylphosphine oxide ≥98%
Cyclohexyldiphenylphosphine oxide
Purity: 98+% CAS No.: 13689-20-8 Molecular Formula: C18H21OP Formula Weight: 284.34 Melting Point: 166-167° Beilstein Reference: 2943737
Formula: C₁₈H₂₁OP
MW: 284.34 g/mol
Melting Pt: 166…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00014301
CAS Number: 13689-20-8
EINECS: 237-204-6