4-Nitro-1,8-naphthalic anhydride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6-Nitrobenzo[de]isochromene-1,3-dione

M-Clarity™ quality level = MQ100

324345-25G 324345-5G
SIAL324345-25GEA 766 EUR
SIAL324345-25G SIAL324345-5G
4-Nitro-1,8-naphthalic anhydride, Sigma-Aldrich®
4-Nitro-1,8-naphthalic anhydride
Formula: C₁₂H₅NO₅
MW: 243.1718 g/mol
MDL Number: MFCD00013446
CAS Number: 6642-29-1
EINECS: 229-659-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR