2-Amino-5-methyl-4-phenylthiazole
Supplier: Cayman Chemical
13061-100 13061-250 13061-500 13061-1
CAYM13061-100EA 60 EUR
CAYM13061-100 CAYM13061-250 CAYM13061-500 CAYM13061-1
2-Amino-5-methyl-4-phenylthiazole
2-Amino-5-methyl-4-phenylthiazole
Formula: C₁₀H₁₀N₂S
MW: 190.2648 g/mol
MDL Number: MFCD00617257
CAS Number: 30709-67-2