2-(4-Pyridinyl)-4-thiazolecarboxylic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

646032-1G 646032-5G
SIAL646032-1GEA 76.9 EUR
SIAL646032-1G SIAL646032-5G
2-(4-Pyridinyl)-4-thiazolecarboxylic acid, Sigma-Aldrich®
2-(4-Pyridinyl)-4-thiazolecarboxylic acid
Formula: C₉H₆N₂O₂S
MW: 206.22 g/mol
MDL Number: MFCD00171745
CAS Number: 21278-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR