3-Amino-1H-pyrazole-4-carboxylic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Amino-4-pyrazolecarboxylic acid, 3-Aminopyrazole-4-carboxylic acid, 3-Amino-1H-pyrazole-4-carboxylic acid, NSC 89246

A77407-1G A77407-5G
SIALA77407-1GEA 91.9 EUR
SIALA77407-1G SIALA77407-5G
3-Amino-1H-pyrazole-4-carboxylic acid, Sigma-Aldrich®
3-Amino-1H-pyrazole-4-carboxylic acid
Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
MDL Number: MFCD00005239
CAS Number: 41680-34-6
EINECS: 255-493-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR