3-Amino-5-hydroxy-1H-pyrazole
Supplier: Thermo Scientific
Synonyms:
3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one
AC27172EA
AC27172DA
MAYBAC27172EAEA
0
EUR
MAYBAC27172EA
MAYBAC27172DA
3-Amino-5-hydroxy-1H-pyrazole
3-Amino-5-hydroxy-1H-pyrazole
Formula:
C₃H₅N₃O MW: 99.09 g/mol |
MDL Number:
MFCD00022384 CAS Number: 6126-22-3 EINECS: 228-095-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...