3-Bromo-1-(trimethylsilyl)-1-propyne, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane
M-Clarity™ quality level = MQ100
377775-10G 377775-1G
SIAL377775-10GEA 260 EUR
SIAL377775-10G SIAL377775-1G
3-Bromo-1-(trimethylsilyl)-1-propyne, Sigma-Aldrich®
3-Bromo-1-(trimethylsilyl)-1-propyne
Formula: C₆H₁₁BrSi
MW: 191.14 g/mol
MDL Number: MFCD00134460
CAS Number: 38002-45-8