Methyl (R)-(-)-3-hydroxyisobutyrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl (R)-(-)-beta-hydroxyisobutyrate, (R)-Methyl 3-hydroxy-2-methylpropanoate
M-Clarity™ quality level = MQ200
270148-1G 270148-5G
SIAL270148-1GEA 54.3 EUR
SIAL270148-1G SIAL270148-5G
Methyl (R)-(-)-3-hydroxyisobutyrate, Sigma-Aldrich®
Methyl (R)-(-)-3-hydroxyisobutyrate
Formula: C₅H₁₀O₃
MW: 118.1311 g/mol
MDL Number: MFCD00063450
CAS Number: 72657-23-9