tert-Butyl isocyanoacetate, Sigma-Aldrich®

Supplier: Merck

58825-1ML-F 58825-5ML-F
SIAL58825-1ML-FEA 148 EUR
SIAL58825-1ML-F SIAL58825-5ML-F
tert-Butyl isocyanoacetate, Sigma-Aldrich®
tert-Butyl isocyanoacetate
Formula: C₇H₁₁NO₂
MDL Number: MFCD00042952
CAS Number: 2769-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR