Ethyl isocyanoacetate, Sigma-Aldrich®

Supplier: Merck


MQ100

226319-25G 226319-5G
SIAL226319-25GEA 267 EUR
SIAL226319-25G SIAL226319-5G
Ethyl isocyanoacetate, Sigma-Aldrich®
Ethyl isocyanoacetate
Formula: C₅H₇NO₂
MW: 113.12 g/mol
MDL Number: MFCD00000007
CAS Number: 2999-46-4
EINECS: 221-077-9
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR