Methyl isocyanoacetate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

238880-1G 238880-5G
SIAL238880-1GEA 60.1 EUR
SIAL238880-1G SIAL238880-5G
Methyl isocyanoacetate, Sigma-Aldrich®
Methyl isocyanoacetate
Formula: C₄H₅NO₂
MW: 99.09 g/mol
MDL Number: MFCD00000006
CAS Number: 39687-95-1
EINECS: 254-593-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR