4'-(Trifluoromethoxy)acetanilide

Supplier: Thermo Scientific

Synonyms: 4-(Trifluoromethoxy)acetanilide

TL00595.1 TL00595.50
MAYBTL00595.1EA 0 EUR
MAYBTL00595.1 MAYBTL00595.50
4'-(Trifluoromethoxy)acetanilide
4'-(Trifluoromethoxy)acetanilide
Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD00075293
CAS Number: 1737-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR