9-Ethylcarbazole, Sigma-Aldrich®
Supplier: Merck
Synonyms:
9-Ethyl-9H-carbazole, N-Ethylcarbazole
E16600-100G
E16600-500G
SIALE16600-100GEA
55
EUR
SIALE16600-100G
SIALE16600-500G
9-Ethylcarbazole, Sigma-Aldrich®
9-Ethylcarbazole
Formula:
C₁₄H₁₃N MW: 195.27 g/mol |
MDL Number:
MFCD00004967 CAS Number: 86-28-2 EINECS: 201-660-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...