2-Acrylamido-2-methylpropanesulphonic acid sodium salt 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
655821-1L 655821-250ML
SIAL655821-1LEA 205 EUR
SIAL655821-1L SIAL655821-250ML
2-Acrylamido-2-methylpropanesulphonic acid sodium salt 50% in aqueous solution, Sigma-Aldrich®
2-Acrylamido-2-methylpropanesulphonic acid sodium salt
Formula: C₇H₁₂NNaO₄S
MDL Number: MFCD00156481
CAS Number: 5165-97-9
EINECS: 225-948-4