4-Hydroxy-3-methoxyphenylboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

M-Clarity™ quality level = MQ100

518786-1G 518786-5G
SIAL518786-1GEA 66.4 EUR
SIAL518786-1G SIAL518786-5G
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester, Sigma-Aldrich®
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₄
MW: 250.099 g/mol
MDL Number: MFCD02093723
CAS Number: 269410-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR