2-Formyl-4-methoxyphenylboronic acid

Supplier: Thermo Scientific

AC35952.250 AC35952.1
MAYBAC35952.250EA 0 EUR
MAYBAC35952.250 MAYBAC35952.1
2-Formyl-4-methoxyphenylboronic acid
2-Formyl-4-methoxyphenylboronic acid
Formula: C₈H₉BO₄
MW: 179.9657 g/mol
MDL Number: MFCD01319043
CAS Number: 139962-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR