4-Hydroxy-3-methoxybenzonitrile 98
Supplier: Thermo Fisher Scientific
Danger
A18056.14 A18056.22
A18056.14EA 46 EUR
A18056.14 A18056.22
4-Hydroxy-3-methoxybenzonitrile 98
4-Hydroxy-3-methoxybenzonitrile
Purity: 98% CAS No.: 4421-08-3 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Melting Point: 85-89° UN No.: 3439 MDL No.: MFCD00001820
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00001820
CAS Number: 4421-08-3
EINECS: 224-589-0
UN: 3439
ADR: 6.1 III