Methyl-5-aminosalicylate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Methyl 5-amino-2-hydroxybenzoate

M-Clarity™ quality level = MQ100

636436-1G 636436-5G
SIAL636436-1GEA 40.3 EUR
SIAL636436-1G SIAL636436-5G
Methyl-5-aminosalicylate, Sigma-Aldrich®
Methyl 5-aminosalicylate
Formula: C₈H₉NO₃
MW: 167.1620 g/mol
MDL Number: MFCD01317557
CAS Number: 42753-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR