4-Hydroxy-3-(trifluoromethyl)benzonitrile ≥98%
Supplier: Thermo Fisher Scientific
Warning
H31639.03 H31639.06
H31639.03EA 63.4 EUR
H31639.03 H31639.06
4-Hydroxy-3-(trifluoromethyl)benzonitrile ≥98%
4-Hydroxy-3-(trifluoromethyl)benzonitrile
Purity: 98+% CAS No.: 124811-71-8 Molecular Formula: C8H4F3NO Formula Weight: 187.12 Melting Point: 171-175° MDL No.: MFCD06797882
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD06797882
CAS Number: 124811-71-8