Benzo[b]thiophene-3-carboxaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: Benzo[b]thiophene-3-carbaldehyde

494968-1G 494968-5G
SIAL494968-1GEA 27.1 EUR
SIAL494968-1G SIAL494968-5G
Benzo[b]thiophene-3-carboxaldehyde, Sigma-Aldrich®
Benzo[b]thiophene-3-carboxaldehyde
Formula: C₉H₆OS
MW: 162.2083 g/mol
MDL Number: MFCD00052376
CAS Number: 5381-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR