Magnesium hexafluorosilicate hydrate
Supplier: Thermo Fisher Scientific
Danger
89018.36 89018.A3
89018.36EA 68.8 EUR
89018.36 89018.A3
Magnesium hexafluorosilicate hydrate
Magnesium hexafluorosilicate hydrate
Molecular Formula: MgSiF6.xH2O Formula Weight: 166.39(anhy) Physical Form: Powder Density: 1.788 Melting Point: 120° dec. UN No.: 2853 MDL No.: MFCD00016196
Formula: F₆MgSi·H₂O
MW: 166.39 g/mol (anhy)
Melting Pt: 120 °C (dec.)
Density: 1.788
Storage Temperature: Ambient
MDL Number: MFCD00016196
CAS Number: 16949-65-8
EINECS: 241-022-2
UN: 2853
ADR: 6.1 III