2,6-Bis(bromomethyl)naphthalene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

649546-1G 649546-5G
SIAL649546-1GEA 190 EUR
SIAL649546-1G SIAL649546-5G
2,6-Bis(bromomethyl)naphthalene, Sigma-Aldrich®
2,6-Bis(bromomethyl)naphthalene
Formula: C₁₂H₁₀Br₂
MW: 314.0200 g/mol
MDL Number: MFCD00228693
CAS Number: 4542-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR