2-Mercaptopyridine-N-oxide sodium salt, anhydrous 98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium
B20529.03 B20529.06 B20529.14
B20529.03EA 33.8 EUR
B20529.03 B20529.06 B20529.14
2-Mercaptopyridine-N-oxide sodium salt, anhydrous 98%
2-Mercaptopyridine N-oxide sodium salt
Purity: 98% CAS No.: 3811-73-2 Molecular Formula: C5H4NNaOS Formula Weight: 149.15 Melting Point: ca 290° dec. Merck Reference: 14,7994 MDL No.: MFCD01941547
Formula: C₅H₄NNaOS
MW: 149.15 g/mol
Melting Pt: ∼ 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01941547
CAS Number: 3811-73-2
EINECS: 223-296-5
Merck Index: 14,7994