2-Chloro-6-methylnicotinic acid 98+%
Supplier: Thermo Fisher Scientific
270810250 270810050 270811000
ACRO270810250EA 131 EUR
ACRO270810250 ACRO270810050 ACRO270811000
2-Chloro-6-methylnicotinic acid 98+%
2-Chloro-6-methylnicotinic acid
Formula: C₇H₆ClNO₂
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00134165
CAS Number: 30529-70-5
EINECS: 250-229-7
Order Now
Specification Test Results
Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 157 to 161 °C
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %