4,7-Dibromo-2,1,3-benzothiadiazole ≥97%
Supplier: Thermo Scientific
Warning
H56306.14 H56306.06 H56306.03
H56306.14EA 521 EUR
H56306.14 H56306.06 H56306.03
4,7-Dibromo-2,1,3-benzothiadiazole ≥97%
4,7-Dibromo-2,1,3-benzothiadiazole
Formula: C₆H₂Br₂N₂S
MW: 293.97 g/mol
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00658844
CAS Number: 15155-41-6
UN: 2811
ADR: 6.1,III