2,1,3-Benzothiadiazole-5-carbonyl chloride

Supplier: Thermo Scientific

CC09202.250 CC09202.1
MAYBCC09202.250EA 0 EUR
MAYBCC09202.250 MAYBCC09202.1
2,1,3-Benzothiadiazole-5-carbonyl chloride
2,1,3-Benzothiadiazole-5-carbonyl chloride
Formula: C₇H₃ClN₂OS
MDL Number: MFCD02681992
CAS Number: 321309-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR