Bromomethyl methyl ether, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
161756-25G 161756-5G
SIAL161756-25GEA 131 EUR
SIAL161756-25G SIAL161756-5G
Bromomethyl methyl ether, Sigma-Aldrich®
Bromomethyl methyl ether
Formula: C₂H₅BrO
MW: 124.97 g/mol
MDL Number: MFCD00000171
CAS Number: 13057-17-5
UN: 1992
ADR: 3 III