2'-Ethylacetanilide

Supplier: Thermo Scientific

TL00639.1 TL00639.10 TL00639.50
MAYBTL00639.1EA 0 EUR
MAYBTL00639.1 MAYBTL00639.10 MAYBTL00639.50
2'-Ethylacetanilide
2'-Ethylacetanilide
Formula: C₁₀H₁₃NO
CAS Number: 33098-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR