L(+)-α-(4-Hydroxyphenyl)glycine, Sigma-Aldrich®

Supplier: Merck

Synonyms: Oxfenicine, H-Phg(4-OH)-OH

56160-10G 56160-50G
SIAL56160-10GEA 120 EUR
SIAL56160-10G SIAL56160-50G
L(+)-α-(4-Hydroxyphenyl)glycine, Sigma-Aldrich®
L(+)-α-(4-Hydroxyphenyl)glycine
Formula: C₈H₉NO₃
MW: 167.1600 g/mol
MDL Number: MFCD00065932
CAS Number: 32462-30-9
EINECS: 251-061-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR