(±)-3-Amino-3-phenylpropionic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: DL-β-Phenylalanine, 3-Amino-3-phenylpropionic acid, H-DL-β-Phe-OH, β-Aminohydrocinnamic acid

M-Clarity™ quality level = MQ100

159492-25G 159492-5G
SIAL159492-25GEA 141 EUR
SIAL159492-25G SIAL159492-5G
(±)-3-Amino-3-phenylpropionic acid, Sigma-Aldrich®
(±)-3-Amino-3-phenylpropionic acid
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00008064
CAS Number: 614-19-7
EINECS: 210-371-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR