Ruthenium, black ≥99.9% (metals basis)
Supplier: Thermo Scientific
Warning
12354.04 12354.09
12354.04EA 276 EUR
12354.04 12354.09
Ruthenium, black ≥99.9% (metals basis)
Ruthenium
Purity: 99.9% CAS No.: 7440-18-8 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011207
Formula: Ru
MW: 101.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,II
Merck Index: 12,08452