2,4-Dichloro-5-fluorobenzonitrile ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H26464.03
H26464.09
H26464.03EA
53.1
EUR
H26464.03
H26464.09
2,4-Dichloro-5-fluorobenzonitrile ≥97%
2,4-Dichloro-5-fluorobenzonitrile
Purity: 97%
CAS No.: 128593-93-1
Molecular Formula: C7H2Cl2FN
Formula Weight: 190.00
Melting Point: 44-47°
UN No.: 3439
MDL No.: MFCD03840469
Formula:
C₇H₂Cl₂FN Melting Pt: 44…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840469 CAS Number: 128593-93-1 EINECS: None UN: 3439 ADR: 6.1,III |