Homophthalic anhydride 98%
Supplier: Thermo Fisher Scientific
Warning
156300050 156300250 156301000
ACRO156300050EA 32.1 EUR
ACRO156300050 ACRO156300250 ACRO156301000
Homophthalic anhydride 98%
Homophthalic anhydride
Formula: C₉H₆O₃
Melting Pt: 140…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4
Order Now
Specification Test Results
Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %
Phosphate (PO4) ≤0.003 %
Insoluble matter ≤0.05 %