(±)-Aspartic acid
Supplier: EHRENSTORFER GMBH
EHERC10304950EA
50.9
EUR
EHERC10304950
(±)-Aspartic acid
(±)-Aspartic acid
Formula:
C₄H₇NO₄ MW: 133,10 g/mol |
CAS Number:
617-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...