Cerium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
Warning
Synonyms: Cerium(III) chloride hydrate, Cerous chloride
11325.18 11325.30
11325.18EA 67.1 EUR
11325.18 11325.30
Cerium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)
Cerium trichloride hydrate
Cerium(III) chloride hydrate is used as an important raw material the preparation of other cerium salts such as cerium(III) trifluoromethanesulfonate. It is used in the conversion of esters to allylsilanes.
Sensitivity: Air Sensitive and Hygroscopic
Appearance: White crystalline aggregates
Formula: CeCl₃·xH₂O
MW: 246.48 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149633
CAS Number: 19423-76-8
EINECS: 232-227-8
Merck Index: 14,1997