4-Isopropylphenylhydrazine hydrochloride 98
Supplier: Thermo Fisher Scientific
Danger
A12416.03 A12416.06 A12416.14
A12416.03EA 32.2 EUR
A12416.03 A12416.06 A12416.14
4-Isopropylphenylhydrazine hydrochloride 98
4-Isopropylphenylhydrazine hydrochloride
Purity: 98% CAS No.: 118427-29-5 Molecular Formula: C9H14N2.HCl Formula Weight: 186.69 Melting Point: 203° dec. MDL No.: MFCD00013461
Formula: C₉H₁₅ClN₂
MW: 186.69 g/mol
Melting Pt: 203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013461
CAS Number: 118427-29-5