4'-(Trifluoromethyl)acetanilide

Supplier: Thermo Scientific

Synonyms: N-(4-(Trifluoromethyl)phenyl)acetamide, 4'-(Trifluoromethyl)acetanilide, 4-Acetamidobenzotrifluoride, alpha,alpha,alpha-Trifluoro-p-acetotoluidide

BTB07343.1 BTB07343.10 BTB07343.50
MAYBBTB07343.1EA 0 EUR
MAYBBTB07343.1 MAYBBTB07343.10 MAYBBTB07343.50
4'-(Trifluoromethyl)acetanilide
4'-(Trifluoromethyl)acetanilide
Formula: C₉H₈F₃NO
MW: 203.16 g/mol
Melting Pt: 208…211 °C
MDL Number: MFCD00013562
CAS Number: 349-97-3
EINECS: 206-495-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR