MES sodium salt ≥98%, Reagent Grade

Supplier: VWR Chemicals

Synonyms: MES-Na

X218-500GEA 661 EUR
X218-500G
MES sodium salt ≥98%, Reagent Grade
MES sodium salt

Order Now

Specification Test Results

Appearance White crystalline powder
Moisture ≤10%
pH (1% Water) @ 25 °C 9 − 10.5
Purity ≥98%
Solubility (10%, Water) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR