Calcium chloride 1 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
21115-100ML 21115-250ML
SIAL21115-100MLEA 75.5 EUR
SIAL21115-100ML SIAL21115-250ML
Calcium chloride 1 mol/l in aqueous solution, Sigma-Aldrich®
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Merck Index: 14,01659