Sodium fluoride 99,99 Metals basis
Supplier: Thermo Fisher Scientific
Danger
46912.09 46912.18
46912.09EA 82 EUR
46912.09 46912.18
Sodium fluoride 99,99 Metals basis
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1704 °C
Melting Pt: 993
Density: 2.78
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 14,8618
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM