4'-Isopropylacetanilide
Supplier: Thermo Scientific
Synonyms: 4-Isopropyl acetanilide
DP01941.1 DP01941.10 DP01941.50
MAYBDP01941.1EA 0 EUR
MAYBDP01941.1 MAYBDP01941.10 MAYBDP01941.50
4'-Isopropylacetanilide
4'-Isopropylacetanilide
Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
MDL Number: MFCD00026406
CAS Number: 5702-74-9