HPLC columns, GlycanPac™ AXH-1
Supplier: Thermo Fisher Scientific
082468 082469 082470 082472 082473 082474 082475 082476 082521
555-7539EA 1330 EUR
555-7539 555-7540 555-7541 555-7459 555-7363 555-7364 555-7365 555-7366 555-7371
HPLC columns, GlycanPac™ AXH-1
Chromatography Columns
These columns are high performance, silica-based HPLC columns for simultaneous separation of glycans by charge, size and polarity. They are designed for high-resolution and high-throughput analysis with unique selectivity for biologically important glycans, either labelled or native, by LC-fluorescence and LC-MS methods.
  • Useful for both high-resolution glycan profile characterisation and easy quantification of glycans based on charge
  • High chromatographic efficiency and excellent column stability
Particle type: high purity, spherical, porous silica
Max. pressure: 6000 psi for 3 µm columns; 7000 psi for 1,9 µm 2,1×100 mm columns; 10000 psi for 1,9 µm 2,1×150 mm columns; 15000 psi for 1,9 µm 2,1×250 mm columns
Flow rate range: 0,6 to 1,2 ml/min for 4,6 mm ID formats; 0,3 to 0,6 ml/min for 3,0 mm ID formats; 0,1 to 0,4 ml/min for 2,1 mm ID formats
pH range: 2,0 to 8,0
Max. temperature: 60 °C