VWR® Oslo, Safety Spectacles
Supplier: VWR
Oslo
111-0932EA 6 EUR
111-0932
VWR® Oslo, Safety Spectacles
Safety Glasses
Safety spectacles with wrap-around, clear polycarbonate lens and indirect ventilation.
  • Anti-scratch/anti-fog coating
  • Integrated side shields
  • Integrated brow guard
  • Adjustable temple length
Weight: 35 g
Certifikace: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)