Safety spectacles, Hustler
Supplier: Bollé Safety
HUSTFLASH
111-1004EA 79.2 EUR
111-1004
Safety spectacles, Hustler
Safety Glasses
Ultra-wrap and sporty, these safety spectacles perfectly combine comfort, protection and design. Available in smoke and blue flash polarised version.
  • Side protection
  • Non slip bridge
  • Anti scratch, anti fog coatings
  • Non slip temples
Weight: 25 g
Certifikace: EN 160, EN 170, EN 172