uvex i-3 Safety Spectacles
Supplier: UVEX
9190.275 9190.280 6108.211 9190.175 6108.210 9190.281 9190.880 9190.885 9190.839 9190.838
111-1239EA 40 EUR
111-1239 111-1669 111-1749 UVEX9190.175 111-1757 111-1874 111-1875 UVEX9190.885 UVEX9190.839 UVEX9190.838
uvex i-3 Safety Spectacles
Safety Glasses
These innovative safety spectacles are designed for a perfect fit.
  • Flexible side arms with variable inclination
  • Integrated Softflex zone ensures comfort for all head sizes
  • Soft nosepiece, earpieces and Softflex zone at temples ensure a comfortable and secure fit
  • Model i-3 add: Prescription spectacles with integrated dioptre correction for near vision
Certifikace: EN 166, EN 170.