Full face mask, X-plore® 6530
Supplier: Dräger
X-plore®
111-1058EA 300 EUR
111-1057 111-1058
Full face mask, X-plore® 6530
Respirators Reusable Respirators Full Face Respirators
The X-plore® 6530 model features breathing filters from the extensive Dräger X-plore® Rd40 filter range and so sets itself apart through being suitable for use in a host of applications.
  • Sealing frame with threefold sealing edge offers reliable protection and a seal-ensuring fit for all facial shapes
  • Comfortable five-point harness makes the product quick to put on and prevents pressure points from forming
  • Distortion-free visor with 180° angle guarantees a large field of vision with a wide, panoramic view
  • Visor made from scratch and chemical resistant Triplex with ultra-stable stainless steel frame (type: 6530 EPDM/Triplex)
  • Intelligent ventilation system with separate inner mask ensures the visor does not become fogged and guarantees an unobstructed view
  • Barcode on the inside makes it easy to record and maintain the mask inventory
  • Integrated stainless steel speech membrane facilitates communication
Certifikace: EN 136 Class 3 (CE marking), NIOSH (PC), AS/NZS
Upozornení: *One size fits all