Chemical Resistant Overalls, 4535
Supplier: 3M
45353XL 45352XL 4535L 4535S
113-6296EA 294 EUR
113-6296 113-6295 113-6297 113-6299
Chemical Resistant Overalls, 4535
Disposable Garments
These disposable overalls made of PP with a PE laminate provide an excellent barrier against dry particles and certain limited liquid chemical splashes.
  • Elasticated waist and ankles for freedom of movement and knitted cuffs for extra comfort
  • Extra large breathable back panel (SMMMS) for reduced heat stress and increased comfort
  • Two-way zip, hood and sealable storm flap for enhanced protection
  • Low linting properties and anti-static coating
Typical light duty applications may include paint spraying, industrial clean-up and maintenance, industrial and chemical processing, building cleaning, insulation laying, pesticide handling/agriculture (particulate aerosols only), powder coating and pharmaceutical manufacturing.
Colour: White/blue
Certifikace: Type 5/6, EN 1149-5, EN 1073-2:2002; Cat. III.