trans-4-(Aminomethyl)cyclohexanecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
857653-10G 857653-250G 857653-50G
SIAL857653-10GEA 64 EUR
SIAL857653-10G SIAL857653-250G SIAL857653-50G
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, Sigma-Aldrich®
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Formula: C₈H₁₅NO₂
MDL Number: MFCD00001466
CAS Number: 1197-18-8
EINECS: 214-818-2