1,1'-Azobis(N,N-dimethylformamide) 95%
Supplier: Thermo Scientific
J64865.03 J64865.06
J64865.03EA 199 EUR
J64865.03 J64865.06
1,1'-Azobis(N,N-dimethylformamide) 95%
1,1'-Azobis(N,N-dimethylformamide)
Formula: C₆H₁₂N₄O₂
Melting Pt: 112 °C
Storage Temperature: Freezer
MDL Number: MFCD00008318
CAS Number: 10465-78-8
EINECS: 233-951-7