2-(Trifluoromethyl)acrylic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(Trifluoromethyl)propenoic acid

M-Clarity™ quality level = MQ100

369144-1G 369144-5G
SIAL369144-1GEA 102 EUR
SIAL369144-1G SIAL369144-5G
2-(Trifluoromethyl)acrylic acid, Sigma-Aldrich®
2-(Trifluoromethyl)acrylic acid
Formula: C₄H₃F₃O₂
MW: 140.06 g/mol
MDL Number: MFCD00042424
CAS Number: 381-98-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR